accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15721-02-5

IUPAC Name: 2,2',5,5'-tetrachloro-[1,1'-biphenyl]-4,4'-diamine
Synonyms: 2,2',5,5'-tetrachloro-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 322.01
Molecular Formula: C12H8Cl4N2
InChi Key: UXOXUHMFQZEAFR-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(C=C1Cl)C1=CC(Cl)=C(N)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results