Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',5,5'-Tetrachlorobenzidine 98.0+%, TCI America™
SDP

Catalog No. T0933500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0933500G 500 g
T093325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0933500G Supplier TCI America Supplier No. T0933500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15721-02-5
Molecular Formula C12H8Cl4N2
Molecular Weight (g/mol) 322.01
MDL Number MFCD00043918
InChI Key UXOXUHMFQZEAFR-UHFFFAOYSA-N
PubChem CID 27465
ChEBI CHEBI:82540
IUPAC Name 2,2',5,5'-tetrachloro-[1,1'-biphenyl]-4,4'-diamine
SMILES NC1=CC(Cl)=C(C=C1Cl)C1=CC(Cl)=C(N)C=C1Cl

Specifications

Melting Point 140°C
Quantity 500 g
Synonym 4,4′-Diamino-2,2′,5,5′-tetrachlorobiphenyl
Formula Weight 322.01
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′,5,5′-Tetrachlorobenzidine
EINECSNumber (4)-0804
RTECSNumber DD1751000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.