accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157230-67-6

IUPAC Name: 2-(diethoxyphosphoryl)-2-methyl-3,4-dihydro-2H-pyrrol-1-ium-1-olate
Synonyms: 2-(diethoxyphosphoryl)-2-methyl-3,4-dihydropyrrol-1-ium-1-olate
Molecular Weight (g/mol): 235.22
Molecular Formula: C9H18NO4P
InChi Key: OKCDBZSDRSXFIB-UHFFFAOYNA-N
SMILES: CCOP(=O)(OCC)C1(C)CCC=[N+]1[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results