Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Diethylphosphono)-5-methyl-1-pyrroline N-Oxide, TCI America™
SDP

Catalog No. D306850MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D306850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D306850MG Supplier TCI America Supplier No. D306850MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 157230-67-6
Molecular Formula C9H18NO4P
Molecular Weight (g/mol) 235.22
InChI Key OKCDBZSDRSXFIB-UHFFFAOYSA-N
Synonym DEPMPO
PubChem CID 119606
IUPAC Name 2-diethoxyphosphoryl-2-methyl-1-oxido-3,4-dihydropyrrol-1-ium
SMILES CCOP(=O)(C1(CCC=[N+]1[O-])C)OCC

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 235.22
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 5-(Diethylphosphono)-5-methyl-1-pyrroline N-Oxide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.