accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSnH3CCH3NH3C

CAS RN 157427-46-8

IUPAC Name: 1-methyl-2-(tributylstannyl)-1H-indole
Synonyms: 1-methyl-2-(tributylstannyl)indole
Molecular Weight (g/mol): 420.23
Molecular Formula: C21H35NSn
InChi Key: QHCNIBQIQQKJPD-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC2=CC=CC=C2N1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results