accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CFFFFFCl

CAS RN 157499-19-9

IUPAC Name: chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
Synonyms: chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
Molecular Weight (g/mol): 302.74
Molecular Formula: C11H12ClF5Si
InChi Key: OCIDTPKJLONLEN-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCCC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results