Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane 95.0+%, TCI America™
SDP

Catalog No. C27005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27005G Supplier TCI America Supplier No. C27005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157499-19-9
Molecular Formula C11H12ClF5Si
Molecular Weight (g/mol) 302.744
MDL Number MFCD00054911
InChI Key OCIDTPKJLONLEN-UHFFFAOYSA-N
Synonym Dimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silyl Chloride, 1-[3-(Chlorodimethylsilyl)propyl]-2,3,4,5,6-pentafluorobenzene
PubChem CID 2760297
IUPAC Name chloro-dimethyl-[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
SMILES C[Si](C)(CCCC1=C(C(=C(C(=C1F)F)F)F)F)Cl

Specifications

Color Yellow
Boiling Point 73°C
Quantity 5 g
UN Number 2987
Formula Weight 302.74
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.