CAS RN 157528-56-8
CAS RN 157528-56-8
IUPAC Name:
1-benzylpyrrolidine-3-carbonitrile
Synonyms:
1-benzylpyrrolidine-3-carbonitrile
Molecular Weight (g/mol):
186.26
Molecular Formula:
C12H14N2
InChi Key:
RYCQUUNQHVAFSM-UHFFFAOYNA-N
SMILES:
N#CC1CCN(CC2=CC=CC=C2)C1
1
–
1
of
1
results
1
(R)-1-Benzyl-3-pyrrolidinecarbonitrile 98.0+%, TCI America™
CAS: 157528-56-8 Molecular Formula: C12H14N2 Molecular Weight (g/mol): 186.258 MDL Number: MFCD09951967 InChI Key: RYCQUUNQHVAFSM-LBPRGKRZSA-N Synonym: (R)-1-Benzyl-3-cyanopyrrolidine PubChem CID: 1515497 IUPAC Name: (3R)-1-benzylpyrrolidine-3-carbonitrile SMILES: C1CN(CC1C#N)CC2=CC=CC=C2