accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 157585-49-4

IUPAC Name: (2,6-dimethylnaphthalen-1-yl)[6-(2,6-dimethylnaphthalene-1-carbonyl)naphthalen-2-yl]methanone
Synonyms: (2,6-dimethylnaphthalen-1-yl)[6-(2,6-dimethylnaphthalene-1-carbonyl)naphthalen-2-yl]methanone
Molecular Weight (g/mol): 492.62
Molecular Formula: C36H28O2
InChi Key: JWNNRHQJCSQBPN-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=C(C)C(C(=O)C3=CC4=CC=C(C=C4C=C3)C(=O)C3=C(C)C=CC4=CC(C)=CC=C34)=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results