Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(2,6-dimethyl-1-naphthoyl)naphthalene, TCI America™
SDP

Catalog No. B19151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19151G Supplier TCI America Supplier No. B19151G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 157585-49-4
Molecular Formula C36H28O2
Molecular Weight (g/mol) 492.618
MDL Number MFCD01321142
InChI Key JWNNRHQJCSQBPN-UHFFFAOYSA-N
PubChem CID 44630147
IUPAC Name [6-(2,6-dimethylnaphthalene-1-carbonyl)naphthalen-2-yl]-(2,6-dimethylnaphthalen-1-yl)methanone
SMILES CC1=CC2=C(C=C1)C(=C(C=C2)C)C(=O)C3=CC4=C(C=C3)C=C(C=C4)C(=O)C5=C(C=CC6=C5C=CC(=C6)C)C

Specifications

Melting Point 260°C
Color Yellow
Quantity 1 g
Formula Weight 492.62
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(2,6-dimethyl-1-naphthoyl)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.