accessibility menu, dialog, popup

UserName

Viewing profile as user:
KOHOHSHOOOSOHOO

CAS RN 15763-57-2

IUPAC Name: 2,5-dihydroxybenzene-1,4-disulfonic acid potassium
Synonyms: 2,5-dihydroxybenzene-1,4-disulfonic acid potassium
Molecular Weight (g/mol): 309.32
Molecular Formula: C6H6KO8S2
InChi Key: TYGULVOTNMOPMK-UHFFFAOYSA-N
SMILES: [K].OC1=CC(=C(O)C=C1S(O)(=O)=O)S(O)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results