Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxy-1,4-benzenedisulfonic Acid,dipotassium Salt 98%, Thermo Scientific™

Catalog No. AC171101000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171101000 100 g
AC171100250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171101000 Supplier Thermo Scientific Chemicals Supplier No. 171101000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15763-57-2
Molecular Formula C6H6K2O8S2
Molecular Weight (g/mol) 348.423
InChI Key IGDNXDWYQNPZMG-UHFFFAOYSA-N
Synonym hydroquinone-2,5-disulfonic acid dipotassium salt
PubChem CID 86591117
IUPAC Name 2,5-dihydroxybenzene-1,4-disulfonic acid;potassium
SMILES C1=C(C(=CC(=C1S(=O)(=O)O)O)S(=O)(=O)O)O.[K].[K]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Linear Formula (HO)2C6H2(SO3K)2
Quantity 100 g
Beilstein 11, 300
Formula Weight 346.4
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2,5-Dihydroxy-1,4-benzenedisulfonic acid,dipotassium salt, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 239-853-0
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.