accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1577-19-1

IUPAC Name: (3E)-oct-3-enoic acid
Synonyms: trans-3-octenoic acid
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: IWPOSDLLFZKGOW-AATRIKPKSA-N
SMILES: CCCC\C=C\CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results