Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octenoic Acid 95.0+%, TCI America™
SDP

Catalog No. O00705ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00705ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O00705ML Supplier TCI America Supplier No. O00705ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1577-19-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00016562
InChI Key IWPOSDLLFZKGOW-AATRIKPKSA-N
PubChem CID 5282716
IUPAC Name (3E)-oct-3-enoic acid
SMILES CCCC\C=C\CC(O)=O

Specifications

Color Yellow
Quantity 5 mL
UN Number 3265
Formula Weight 142.20
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Octenoic Acid
EINECSNumber (2)-0609
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.