CAS RN 157771-56-7
IUPAC Name:
2,7-dibromo-9,9-dipropyl-9H-fluorene
Synonyms:
2,7-dibromo-9,9-dipropylfluorene
Molecular Weight (g/mol):
408.18
Molecular Formula:
C19H20Br2
InChi Key:
QQTBEFPDRHYPLE-UHFFFAOYSA-N
SMILES:
CCCC1(CCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
1
–
1
of
1
results
1
2,7-Dibromo-9,9-dipropylfluorene 98.0+%, TCI America™
CAS: 157771-56-7 Molecular Formula: C19H20Br2 Molecular Weight (g/mol): 408.18 MDL Number: MFCD12024280 InChI Key: QQTBEFPDRHYPLE-UHFFFAOYSA-N PubChem CID: 21924531 IUPAC Name: 2,7-dibromo-9,9-dipropyl-9H-fluorene SMILES: CCCC1(CCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2