Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9-dipropylfluorene 98.0+%, TCI America™
SDP

Catalog No. D45695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45695G 5 g
D45691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45695G Supplier TCI America Supplier No. D45695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 157771-56-7
Molecular Formula C19H20Br2
Molecular Weight (g/mol) 408.18
MDL Number MFCD12024280
InChI Key QQTBEFPDRHYPLE-UHFFFAOYSA-N
PubChem CID 21924531
IUPAC Name 2,7-dibromo-9,9-dipropyl-9H-fluorene
SMILES CCCC1(CCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 139°C
Quantity 5 g
Formula Weight 408.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-dipropylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.