accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOOOOHONHNNHH2NH3CH3CCH3H3CCH3(R)(E)

CAS RN 157774-30-6

IUPAC Name: (2R)-5-[(E)-[amino(2,2,5,7,8-pentamethyl-3,4-dihydro-2H-1-benzopyran-6-sulfonamido)methylidene]amino]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: (2R)-5-[(E)-[amino(2,2,5,7,8-pentamethyl-3,4-dihydro-1-benzopyran-6-sulfonamido)methylidene]amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 662.8
Molecular Formula: C35H42N4O7S
InChi Key: QTWZCODKTSUZJN-GDLZYMKVSA-N
SMILES: CC1=C(C)C(=C(C)C2=C1OC(C)(C)CC2)S(=O)(=O)N\C(N)=N\CCC[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)