accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNH2OOOHHO

CAS RN 157798-13-5

IUPAC Name: 2-[5-(nonyloxy)-1H-indol-3-yl]ethan-1-amine; oxalic acid
Synonyms: 5-nonyloxytryptamine oxalate O-nonylserotonin oxalate
Molecular Weight (g/mol): 392.5
Molecular Formula: C21H32N2O5
InChi Key: JORSCLBFSAAOFR-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CCCCCCCCCOC1=CC=C2NC=C(CCN)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results