accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NON

CAS RN 157837-31-5

IUPAC Name: 3-(1,3-oxazol-5-yl)aniline
Synonyms: 3-(1,3-oxazol-5-yl)aniline
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: AIELNJDAOGTASK-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results