Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(1,3-oxazol-5-yl)aniline

Catalog No. CDS014206 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014206 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014206 Supplier Sigma Aldrich Supplier No. CDS014206250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is an aryl halide. it can be obtained from 5-bromo-m-xylene, via copper-catalyzed halogen exchange reaction, in the presence of nai or ki in n-buoh or dmf (solvents). it undergoes reaction with phenol in the presence of cufe2o4 nano powder as a recyclable catalyst to afford 1,3-dimethyl-5-phenoxybenzene.1-iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of n-(3,5-xylyl)-n-ethylaniline, an arylamine.it may be used in the following studies:
  • α-arylation of ketones
  • Copper-catalyzed n-arylation of imidazoles
  • Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile
  • Synthesis of 1,3-dimethyl-5-phenoxybenzene by nano-cufe2o4 catalyzed c-o cross-coupling with phenol
  • Cubr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form n-allyl-3,5-dimethylbenzenamine
  • Copper-catalyzed c-s bond-formation between 5-iodo-m-xylene and thiophenol
  • As a starting material in the synthesis of biphenyl-3,3',5,5'-tetracarboxylic acid
  • Radical bromination of 5-iodo-m-xylene by n-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene

Specifications

CAS 157837-31-5
Molecular Formula C9H8N2O
Linear Formula C9H8N2O
MDL Number MFCD00052191
Quantity 25 mL
Molecular Weight (g/mol) 160.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.