accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOBr

CAS RN 157893-14-6

IUPAC Name: 3-bromo-5-methoxybenzoic acid
Synonyms: 3-bromo-5-methoxybenzoic acid
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: DMXJBCHYVUGXEH-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results