accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONClFFFFF

CAS RN 1580464-71-6

IUPAC Name: methyl 2-chloro-4-(1,1,2,2,2-pentafluoroethyl)pyridine-3-carboxylate
Synonyms: methyl 2-chloro-4-(1,1,2,2,2-pentafluoroethyl)pyridine-3-carboxylate
Molecular Weight (g/mol): 289.59
Molecular Formula: C9H5ClF5NO2
InChi Key: GUORMFCKJWQHIU-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CN=C1Cl)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results