accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOIBrOO

CAS RN 1580548-81-7

IUPAC Name: 5-bromo-3-oxo-3H-1λ³,2-benziodaoxol-1-yl acetate
Synonyms: 5-bromo-3-oxo-1λ³,2-benziodaoxol-1-yl acetate
Molecular Weight (g/mol): 384.95
Molecular Formula: C9H6BrIO4
InChi Key: QPZPQFBDNWCSGA-UHFFFAOYSA-N
SMILES: CC(=O)O[I]1OC(=O)C2=C1C=CC(Br)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results