Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetoxy-5-bromo-1,2-benziodoxol-3(1H)-one 97.0+%, TCI America™
SDP

Catalog No. A26781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26781G Supplier TCI America Supplier No. A26781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1580548-81-7
Molecular Formula C9H6BrIO4
Molecular Weight (g/mol) 384.951
InChI Key QPZPQFBDNWCSGA-UHFFFAOYSA-N
Synonym ABBX
PubChem CID 91972141
IUPAC Name (5-bromo-3-oxo-1$l^{3},2-benziodoxol-1-yl) acetate
SMILES CC(=O)OI1C2=C(C=C(C=C2)Br)C(=O)O1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 384.95
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Acetoxy-5-bromo-1,2-benziodoxol-3(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.