accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 158098-50-1

IUPAC Name: (naphthalen-1-yl)[4-(2-phenylethyl)phenyl]methanone
Synonyms: (naphthalen-1-yl)[4-(2-phenylethyl)phenyl]methanone
Molecular Weight (g/mol): 336.43
Molecular Formula: C25H20O
InChi Key: CFQMRXPGNSGVOV-UHFFFAOYSA-N
SMILES: O=C(C1=CC=C(CCC2=CC=CC=C2)C=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results