Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dibenzyl 1-Naphthyl Ketone 98.0+%, TCI America™
SDP

Catalog No. D22771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22771G Supplier TCI America Supplier No. D22771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 158098-50-1
Molecular Formula C25H20O
Molecular Weight (g/mol) 336.434
MDL Number MFCD00191634
InChI Key CFQMRXPGNSGVOV-UHFFFAOYSA-N
Synonym 4-(1-Naphthoyl)dibenzyl, 4′C-(1-Naphthoyl)-1,2-diphenylethane
PubChem CID 44630255
IUPAC Name naphthalen-1-yl-[4-(2-phenylethyl)phenyl]methanone
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)C(=O)C3=CC=CC4=CC=CC=C43

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
Formula Weight 336.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dibenzyl 1-Naphthyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.