accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NOOFFFNOO

CAS RN 1582-09-8

IUPAC Name: 2,6-dinitro-N,N-dipropyl-4-(trifluoromethyl)aniline
Synonyms: tristar trifluralin (iso) trifluralin
Molecular Weight (g/mol): 335.28
Molecular Formula: C13H16F3N3O4
InChi Key: ZSDSQXJSNMTJDA-UHFFFAOYSA-N
SMILES: CCCN(CCC)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results