Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluralin, Restek
SDP

Catalog No. 06719864 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-864 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-864 Supplier Restek Corporation Supplier No. 32238
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1582-09-8
Molecular Formula C13H16F3N3O4
Molecular Weight (g/mol) 335.283
InChI Key ZSDSQXJSNMTJDA-UHFFFAOYSA-N
PubChem CID 5569
ChEBI CHEBI:35027
IUPAC Name 2,6-dinitro-N,N-dipropyl-4-(trifluoromethyl)aniline
SMILES CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-]

Specifications

Quantity 1 mL
Chemical Name or Material Trifluralin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.