accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1582-27-0

IUPAC Name: 1-(4-chlorophenyl)diazyne-1,2-diium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-(4-chlorophenyl)diazyne-1,2-diium hexafluorophosphate
Molecular Weight (g/mol): 285.53
Molecular Formula: C6H5ClF6N2P
InChi Key: GPRWHKMKUTZAGG-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.ClC1=CC=C(C=C1)[N+]#[NH+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results