Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzenediazonium Hexafluorophosphate, Thermo Scientific™

Catalog No. AC175150500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175150500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175150500 Supplier Thermo Scientific Chemicals Supplier No. 175150500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1582-27-0
Molecular Formula C6H4ClF6N2P
Molecular Weight (g/mol) 284.526
InChI Key JOLVAGIWTVZCNF-UHFFFAOYSA-N
Synonym 4-chlorobenzenediazonium hexafluorophosphate, phosfluorogen a, 4-chlorobenzenediazonium hexafluorophosphate 1-, benzenediazonium, 4-chloro-, hexafluorophosphate 1-1:1, p-chlorobenzenediazonium hexa-fluorophosphate, 4-chlorobenzene-diazonium hexafluorophosphate
PubChem CID 2723983
IUPAC Name 4-chlorobenzenediazonium;hexafluorophosphate
SMILES C1=CC(=CC=C1[N+]#N)Cl.F[P-](F)(F)(F)(F)F

Specifications

Quantity 50 g
Linear Formula ClC6H4N2PF6
Formula Weight 284.52
Chemical Name or Material 4-Chlorobenzenediazonium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.