accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFF

CAS RN 1583-83-1

IUPAC Name: 1-(difluoromethoxy)-4-methylbenzene
Synonyms: 1-(difluoromethoxy)-4-methylbenzene
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: DJDQNISEJVPQCS-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results