Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Difluoromethoxy)toluene 96.0+%, TCI America™
SDP

Catalog No. D49361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D49361G Supplier TCI America Supplier No. D49361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1583-83-1
Molecular Formula C8H8F2O
Molecular Weight (g/mol) 158.148
MDL Number MFCD00042253
InChI Key DJDQNISEJVPQCS-UHFFFAOYSA-N
Synonym alpha,alpha-Difluoro-4-methylanisole
PubChem CID 74104
IUPAC Name 1-(difluoromethoxy)-4-methylbenzene
SMILES CC1=CC=C(C=C1)OC(F)F

Specifications

Color Yellow
Boiling Point 167°C
Quantity 1 g
Formula Weight 158.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Difluoromethoxy)toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.