accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOOH2OH2OH2OCa

CAS RN 15842-29-2

IUPAC Name: calcium trihydrate dinitrate
Synonyms: calcium trihydrate dinitrate
Molecular Weight (g/mol): 218.13
Molecular Formula: CaH6N2O9
InChi Key: BQOJVQYLMGADDA-UHFFFAOYSA-N
SMILES: O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results