Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Nitrate Trihydrate 99.999%, Thermo Scientific™

Catalog No. AC192750200
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC192750200 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC192750200 Supplier Thermo Scientific Chemicals Supplier No. 192750200

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15842-29-2
Molecular Formula CaH6N2O9
Molecular Weight (g/mol) 218.131
InChI Key BQOJVQYLMGADDA-UHFFFAOYSA-N
Synonym calcium nitrate trihydrate, calcium trihydrate dinitrate, ca.2no3.3h2o, calcium nitrate trihydrate., calcium nitrate-water 1/2/3, nitric acid, calciumsalt, trihydrate 8ci,9ci
PubChem CID 57370235
IUPAC Name calcium;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Ca+2]

Specifications

Quantity 20 g
Linear Formula Ca(NO3)2.3H2O
Merck Index 10, 1659
Formula Weight 218.14
Chemical Name or Material Calcium nitrate trihydrate, 99.999%
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.