accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 158478-81-0

OOOHOOOOONHNHNNH(S)H3CH3CCH3CH3CH3H3C

CAS RN 158478-81-0

IUPAC Name: (2S)-6-{[bis({[(tert-butoxy)carbonyl]amino})methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-({bis[(tert-butoxycarbonyl)amino]methylidene}amino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 610.71
Molecular Formula: C32H42N4O8
InChi Key: MIEGDMIQQQRLOZ-VWLOTQADSA-N
SMILES: CC(C)(C)OC(=O)NC(NC(=O)OC(C)(C)C)=NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)