accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15851-44-2

IUPAC Name: cadmium(2+) tellurite
Synonyms: cadmium(2+) tellurite
Molecular Weight (g/mol): 288.01
Molecular Formula: CdO3Te
InChi Key: GKMPTXZNGKKTDU-UHFFFAOYSA-L
SMILES: [Cd++].[O-][Te]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results