Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium tellurite, 99%

Catalog No. AAA1747614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1747614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1747614 Supplier Thermo Scientific Chemicals Supplier No. A1747614
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15851-44-2 | CdO3Te | 288.01 g/mol

It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15851-44-2
Molecular Formula CdO3Te
Molecular Weight (g/mol) 288.01
MDL Number MFCD00049413
InChI Key GKMPTXZNGKKTDU-UHFFFAOYSA-L
Synonym cadmium tellurite, cadmium 2+ tellurite, cadmium tellurium trioxide, cd.o3te, acmc-20alj5, cadmium 2+ ion tellurite, telluric acid h2teo3 , cadmium salt 1:1, tellurous acid cadmium salt, telluric acid h2teo3 ,cadmium salt 1:1
PubChem CID 167475
IUPAC Name cadmium(2+);tellurite
SMILES [Cd++].[O-][Te]([O-])=O

Specifications

Quantity 25 g
UN Number UN2570
Solubility Information Insoluble in water.
Formula Weight 288.01
Percent Purity 99%
Chemical Name or Material Cadmium tellurite
Hazard Category H302+H312+H332-H350
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 239-963-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.