accessibility menu, dialog, popup

UserName

Viewing profile as user:
CClSbClClClClCl

CAS RN 1586-91-0

IUPAC Name: hexachlorostibanuide; triphenylmethylium
Synonyms: hexachlorostibanuide; triphenylmethylium
Molecular Weight (g/mol): 577.79
Molecular Formula: C19H15Cl6Sb
InChi Key: GASWLBDVMZXHOP-UHFFFAOYSA-H
SMILES: Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results