CAS RN 158681-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ SR 141716A
A Tocris product, part of R&D Systems. Selective CB1 inverse agonist