accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15869-85-9

IUPAC Name: 5-methylnonane
Synonyms: nonane, 5-methyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: TYSIILFJZXHVPU-UHFFFAOYSA-N
SMILES: CCCCC(C)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results