Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylnonane 99.0+%, TCI America™
SDP

Catalog No. M02845ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02845ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02845ML Supplier TCI America Supplier No. M02845ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15869-85-9
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
MDL Number MFCD00048832
InChI Key TYSIILFJZXHVPU-UHFFFAOYSA-N
PubChem CID 27518
IUPAC Name 5-methylnonane
SMILES CCCCC(C)CCCC

Specifications

Color Colorless
Boiling Point 165°C
Quantity 5 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methylnonane
EINECSNumber (2)-0010
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.