accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3C

CAS RN 15869-95-1

IUPAC Name: 4,4-dimethyloctane
Synonyms: 4,4-dimethyloctane
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: ZMEDGZAGMLTROM-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results