Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyloctane 98.0+%, TCI America™
SDP

Catalog No. D12161ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12161ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12161ML Supplier TCI America Supplier No. D12161ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15869-95-1
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00048813
InChI Key ZMEDGZAGMLTROM-UHFFFAOYSA-N
Synonym 4,4-dimethyl octane, octane, 4,4-dimethyl, acmc-1bqkk
PubChem CID 85151
IUPAC Name 4,4-dimethyloctane
SMILES CCCCC(C)(C)CCC

Specifications

Color Colorless
Boiling Point 161°C
Quantity 1 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyloctane
EINECSNumber (2)-0010
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.