accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOOCH3

CAS RN 1587-15-1

IUPAC Name: 1,4-dimethyl 2-hydroxybutanedioate
Synonyms: 1,4-dimethyl 2-hydroxybutanedioate
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: YSEKNCXYRGKTBJ-UHFFFAOYNA-N
SMILES: COC(=O)CC(O)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results