Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl DL-Malate 97.0+%, TCI America™
SDP

Catalog No. M002510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M002510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M002510G Supplier TCI America Supplier No. M002510G
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1587-15-1
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00025868
InChI Key YSEKNCXYRGKTBJ-UHFFFAOYSA-N
Synonym DL-Apple Acid Dimethyl Ester, DL-Malic Acid Dimethyl Ester
PubChem CID 12674
IUPAC Name dimethyl 2-hydroxybutanedioate
SMILES COC(=O)CC(C(=O)OC)O

Specifications

Color Colorless
Boiling Point 205°C
Quantity 10 g
Formula Weight 162.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl DL-Malate
EINECSNumber (2)-1443
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.