accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1587-21-9

IUPAC Name: 1,2,3-tripropyl 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: 1,2,3-tripropyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol): 318.37
Molecular Formula: C15H26O7
InChi Key: ODHUFJLMXDXVRC-UHFFFAOYSA-N
SMILES: CCCOC(=O)CC(O)(CC(=O)OCCC)C(=O)OCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results