Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropyl Citrate 97.0+%, TCI America™
SDP

Catalog No. C060125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C060125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C060125G Supplier TCI America Supplier No. C060125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1587-21-9
Molecular Formula C15H26O7
Molecular Weight (g/mol) 318.37
MDL Number MFCD00059407
InChI Key ODHUFJLMXDXVRC-UHFFFAOYSA-N
Synonym Citric Acid Tripropyl Ester
PubChem CID 74113
IUPAC Name 1,2,3-tripropyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCCOC(=O)CC(O)(CC(=O)OCCC)C(=O)OCCC

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 318.37
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tripropyl Citrate
EINECSNumber (2)-1320
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.