accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15870-10-7

IUPAC Name: 2-methylhept-1-ene
Synonyms: 2-methyl-1-heptene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: RCBGGJURENJHKV-UHFFFAOYSA-N
SMILES: CCCCCC(C)=C
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)