Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-heptene 98.0+%, TCI America™
SDP

Catalog No. M05345ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05345ML Supplier TCI America Supplier No. M05345ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15870-10-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009517
InChI Key RCBGGJURENJHKV-UHFFFAOYSA-N
Synonym 2-methyl-1-heptene, 1-heptene, 2-methyl, 2-methyl-hept-1-ene, acmc-209dip, rcbggjurenjhkv-uhfffaoysa, 1-heptene, 2-methyl-8ci 9ci
PubChem CID 27519
IUPAC Name 2-methylhept-1-ene
SMILES CCCCCC(=C)C

Specifications

Color Colorless
Boiling Point 119°C
Quantity 5 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-heptene
EINECSNumber (2)-0024
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.