accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15872-43-2

IUPAC Name: 4-(nonyloxy)benzoic acid
Synonyms: 4-(nonyloxy)benzoic acid
Molecular Weight (g/mol): 264.37
Molecular Formula: C16H24O3
InChi Key: BOZLUAUKDKKZHJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results